주요 문서
57463
Rheodyne® Model 3725i Preparative Sample Injector
stainless steel
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
material
stainless steel
특징
manual
number of ports 6
positions 2
포장
pkg of 1 ea
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Model 3725i sample injectors for 1-10 cm I.D. HPLC columns combine the ease of use and versatility of Rheodyne′s analytical-scale injectors with the ability to handle large samples and high flow rates. Rugged and easily maintained, in a choice of stainless steel or PEEK flowpaths. Includes 10 mL sample loop, 1/8 in. fittings for all ports, 5cm large-bore syringe needle (16 gauge/0.65 in. OD) for rapid injection of large samples, needle port cleaner, two vent tubes, wrenches, mounting screws, and instructions.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
Rheodyne is a registered trademark of IDEX Health & Science, LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)