주요 문서
58827
Rheodyne® Model 7010 Injector
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
특징
number of ports 6
positions 2
일반 설명
The original Rheodyne HPLC injector, Model 7010 introduces 5 μL - 5 mL samples onto a column with excellent reproducibility (intended for complete loop filling only). Six clustered ports allow compact interconnections, minimizing tubing volume. The flat-face seal is easy to adjust and service. Operates at pressures to 7000 psi (490 kg/cm2). Supplied with a 20 μL loop; order fill port separately.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
Rheodyne is a registered trademark of IDEX Health & Science, LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)