Principaux documents
EP022822209
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-40-36-12, non aerosol tight, (36 x 1.5 mL flat-bottom tubes)
About This Item
Code UNSPSC :
41103909
Caractéristiques
to hold 36 x 1.5 mL flat-bottom tubes
to hold 36 x 12 x 32 mm tubes
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022822209 (NA)
Technique(s)
centrifugation: suitable
Capacité
(36 x 1.5 mL flat-bottom tubes)
Compatibilité
for use with centrifuge models Vacufuge Plus Concentrator
Description générale
Selection of rotors for the Eppendorf Vacufuge Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG