Principaux documents
RA2VC-EP
Rotor for Eppendorf® Vacufuge® Plus Concentrator
swing-out rotor, A-2-VCm non aerosol tight
About This Item
Code UNSPSC :
41103909
Caractéristiques
skirt
to hold 2 x MTP or skirted PCR plates (max height 27 mm)
to hold 2 x deepwell plates (max height 27 mm)
A-2-VCm non aerosol tight
swing-out rotor
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022822241 (North America)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Vacufuge Plus Concentrator
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG