Principaux documents
CLS5420100
Pyrex® Kjeldahl, round-bottom, long neck flask
capacity 100 mL
Synonyme(s) :
bottom flask, flask round, round bottomed flask, Corning® 5420
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
long
round bottom flask
Taille de l'obturateur
2
Stérilité
non-sterile
Caractéristiques
closure type Rubber stopper
Conditionnement
case of 24
Fabricant/nom de marque
Corning 5420-100
Capacité
100 mL
Taille du col
(1)
Description générale
Flasks, specially styled, have tooled finished necks for extra strength and uniform stopper fit. Wall thickness is controlled to offer the optimum balance between thermal shock and chemical resistance. Can be used in ASTM E-258 procedure. Care must be taken to ensure the bottoms of the flasks do not come in contact with the heating elements. Most Kjeldahl equipment manufacturers have ring suppors for this purpose.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.