Principaux documents
CLS3360500FO
Pyrex® general purpose distilling apparatus, Pyrex stopper, ST joint
stopper and flask only, capacity 500 mL
Synonyme(s) :
Corning® 3360
About This Item
Code UNSPSC :
41121800
Joint
joint stopper27
Fabricant/nom de marque
Corning 3360-500FO
Capacité
500 mL
Description générale
A general purpose batch distillation assembly with the facility or addition via the Pyrex stoppered neck. Ideally suited for the batch production of high purity distilled water. The flask has a flat bottom with a side delivery tube, terminating in a joint. Condenser tubulations are 10mm O.D.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.