Principaux documents
CLS3340500
Pyrex® Kjeldahl nitrogen distilling apparatus, ST joints
capacity 500 mL
Synonyme(s) :
Corning container
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 3340-500
Capacité
500 mL
Catégories apparentées
Description générale
Used for determining nitrogen in organic compounds. This assembly includes a Kjeldahl flask specifically developed for this process. Both the digestion and distillation stages can be performed in this flask. the assembly shown is the distillation stage, where the amount of ammonia, and hence, nitrogen is established. the inlet tube allows the permanganate solution to be introduced after distillation. Supplied with and outer ST 29/42 joint on the flask and an outer ST 19/38 joint on the condenser. Total height is 410mm; total width is 300mm; maximum O.D. is 97 mm.
Informations légales
PYREX is a registered trademark of Corning, Inc.