Principaux documents
CLS215519
Pyrex® distilling column
L 283 mm, joint: ST/NS 19/22
Synonyme(s) :
Corning® 2155
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS19/22
Fabricant/nom de marque
Corning 2155-19
Longueur
283 mm
Catégories apparentées
Description générale
Drip-tip. Simple design, particularly useful in fractionations.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.