Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92100N

Sigma-Aldrich

Probumin® Bovine Serum Albumin Solution(30%)-LS Grade

Solvent Free, pkg of 50 mL

Synonym(s):

BSA Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41141801
NACRES:
NA.25

Quality Level

form

solution

mol wt

~66 kDa

purified by

heat shock fractionation (solvent free)

packaging

pkg of 50 mL

manufacturer/tradename

Probumin®

origin

USA origin

concentration

29-31 g/dL Biuret (protein)

technique(s)

enzyme immunoassay: suitable

impurities

<50 μg/g IgG

pH

6.8-7.4

cation traces

heavy metals: ≤20 ppm

foreign activity

Tested For Mycoplasma
Tested for Microbial Content by USP Method
Tested for Protease

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

A highly monomeric albumin, virtually free of IgG and reduced fatty acids.

Application

Suitable for use in immunoassays.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service