921001
Probumin® Bovine Serum Albumin Solution(30%)-LS Grade
Solvent Free, pkg of 1 L
Synonym(s):
BSA Solution
About This Item
Quality Level
form
solution
mol wt
~66 kDa
purified by
heat shock fractionation (solvent free)
packaging
pkg of 1 L
manufacturer/tradename
Probumin®
origin
USA origin
concentration
29-31 g/dL Biuret (protein)
technique(s)
enzyme immunoassay: suitable
impurities
<50 μg/g IgG
pH
6.8-7.4
cation traces
heavy metals: ≤20 ppm
foreign activity
Tested For Mycoplasma
Tested for Microbial Content by USP Method
Tested for Protease
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service