주요 문서
54632
Rheodyne® Model 9725i PEEK Injector
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
material
PEEK injector
설명
manual; with MBB and switch
특징
manual
number of ports 6
positions 2
제조업체/상표
Rheodyne 9725I
파라미터
5000 psi max. pressure
피팅
10-32 Rheodyne thread
일반 설명
The 9725i contains a switch and has the make before break (MBB). With the MBB, flow is not interupted when switching from laod to inject.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
Rheodyne is a registered trademark of IDEX Health & Science, LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)