주요 문서
29432-U
Supelco
Certified screw thread vials, 10-425 thread, 12 x 32 mm
volume 2 mL, clear glass vial, PTFE/silicone septum, (convenience kit), pkg of 100 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122001
NACRES:
NB.11
material
PTFE/silicone septum
clear glass vial
polypropylene cap
특징
closure type screw top vial
kit with vial, cap, and septa
포장
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 6 mm
용적
2 mL
피팅
thread for 10-425
유사한 제품을 찾으십니까? 방문 제품 비교 안내
기타 정보
The inside diameter of vials vary with the manufacture. The following insert part numbers are compatible with these large opening vials.
  • 29435-UPK100, Certified 200 μL glass inserts, mandrel precision point, 6 X 29mm, with bottom spring
  • 29436-UPK100, Certified 250 μL glass insert, conical bottom, 6 x 31 mm
  • 29437-UPK100, Certified 300 μL glass inserts, conical bottom, 6 x 30 mm
  • 29441-U PK100, Certified 350 μL glass inserts, flat bottom, 6 x 31mm
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)