주요 문서
Z180920
Sigma-Aldrich
Ace auto-zero McLeod vacuum gauges
pressure 0-1 mmHg
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41112406
NACRES:
NB.79
제조업체/상표
Ace Glass 872547
압력 측정 범위
0-1 mmHg
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
A tilting gauge with a constant zero overflow which eliminates the capillary-depression error from an expanding meniscus. Once the zero line of the scale is aligned with the tip of the closed capillary, it is only necessary to bring the gauge to an upright position and read the scale behind the closed leg.
Gauges are fabricated from calibrated ACE Trubore tubing, to assure uniform and reproducible reading between the gauges. Mercury trap is incorporated in female joint, bulbs are enlarged. Joint is standard taper 20/65.
The pressure range is: 0-1.0 mmHg and the lowest reading is: 0.001 mmHg.
Mercury not included (approximately 20 mL of triple distilled mercury is needed to ffill this vacuum gauge to working volume).
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)