주요 문서
Z108510
Sigma-Aldrich
Macro Diazald® kits
joint: ST/NS 24/40 Clear-Seal joints
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104812
NACRES:
NB.43
조인트
joint: ST/NS24/40 Clear-Seal joints
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
For technical and safety information about Diazald and Aldrich diazomethane generators see Technical Information Bulletin AL-180.
For the large-scale (200 to 300 mmol) generation of diazomethane. The dry ice condenser quantitatively condenses all the diazomethane/ether vapor during diazomethane generation from Diazald. Designed by M. Hudlicky [J. Org. Chem., 1980, 45, 5377]. The stopcock is closed at the start of distillation. As the distillate drips off the condenser, the stopcock is opened and the first portions fill the trap, allowing diazomethane/ether condensate to collect in the 500 mL Erlenmeyer flask, but prevents the escape of uncondensed vapors into the receiver. Glassware has Clear-Seal joints, which do not require grease and are safer because of their smooth surfaces (sharp edges and ground glass must be avoided in diazomethane generation). Thermometer not included.
Call or visit our website for pricing on replacement parts.
NOTE: Sigma-Aldrich is unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
법적 정보
Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)