주요 문서
Z104574
Scienceware® flow indicator
gives immediate visual indication
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
20121904
NACRES:
NB.51
포장
pkg of 1 ea
제조업체/상표
Bel-Art H19935-0000
일반 설명
Gives immediate visual indication of cooling-water (or gas) flow by rotation of white ball in the circular channel. A minimum water flow of 50 mL/min (indicator in horizontal position) or 140 mL/min (indicator in vertical position) will actuate the ball. With air, the corresponding minimum flow rates are 3 and 8 L/min. Maximum pressure is 50 psig with water and 10 psig with air.
법적 정보
Scienceware is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)