Principaux documents
Z741974
Cameo Filtration Membrane
Nylon membrane, pore size 5.0 μm, diam. 47 mm, pkg of 100 ea
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Matériaux
Nylon membrane
Stérilité
non-sterile
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
GVS Life Sciences 1213812
Diamètre
47 mm
Dimension de pores
5.0 μm
Description générale
Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.
Applications: Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
Informations légales
Cameo is a trademark of GVS S.p.A.