Principaux documents
Z719536
Sorenson low binding Ultra G microtips
maximum volume 10 μL, pkg of 960 tips QuickRack, non-sterile
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
non-sterile
Caractéristiques
barrier: no
graduations
Conditionnement
case of 4800 ea 50 QuickRack of 96 tips
pack of 960 ea 10 QuickRack of 96 tips
pkg of 960 tips QuickRack
Fabricant/nom de marque
Sorenson 30470T
Volume maximum
10 μL
Volume
10 μL
Description générale
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for ultra-micro pipettors.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.