Principaux documents
Z719447
Sorenson low binding aerosol barrier tips
MultiGuard, maximum volume 200 μL , Graduated
Synonyme(s) :
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
sterile
Caractéristiques
DNase free
RNase free
barrier
graduations
Conditionnement
pack of 96 ea (Racked)
Fabricant/nom de marque
Sorenson 30550T
Volume maximum
200 μL , Graduated
Volume
200 μL
Type de liaison
low binding surface
Description générale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.