Principaux documents
TMO4113-0125
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 125 mL, baffled bottom, case of 24 ea
Synonyme(s) :
Nalgene Erlenmeyer flask, single use plastic flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.13
Matériaux
PETG
Stérilité
sterile
Conditionnement
case of 24 ea
Fabricant/nom de marque
Nalgene 4113-0125
Volume du flacon
125 mL
Description générale
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Autres remarques
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries