T6948
Sartorius pipette tips
volume range 100-5000 μL, standard, bulk packed, non-sterile
Synonyme(s) :
Pipette Tips, Sartorius Pipette Tips, Sartorius Tips
About This Item
Produits recommandés
Matériaux
colorless
Stérilité
non-sterile
Caractéristiques
DNase free
RNase free
barrier: no
endotoxin free
Conditionnement
pack of 100 ea (Color coded tip trays)
Fabricant/nom de marque
Sartorius 780300
Volume maximum
5 mL
Gamme de volumes
100-5000 μL
Volume
100—5000 μL
Catégories apparentées
Description générale
Optifit Tips are available in various packaging types for all your needs:
- Single Tray racks – purity certified tip racks
- Refill Packs - purity certified and ecological refill packages
- Refill Towers - space-saving and ecological refill packages
- Flexibulk boxes - space-saving, ecological and economical bulk boxes
- Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.