Principaux documents
RA438-EP
Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges
A-4-38, aerosol-tight, without buckets, for round-bottom tubes only
About This Item
Code UNSPSC :
41103909
Caractéristiques
to hold 4 x 100 mL round buckets
to hold 4 x 90 mL rectangular buckets
aerosol-tight
for round-bottom tubes only
swing bucket rotor
without buckets
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022639064 (NA)
Eppendorf® 5702740004 (EU)
Paramètres
2,750 × g max. RCF
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5702/5702R/502RH
Description générale
Rotors to fit Eppendorf centrifuge models 5702, 5720 R, and 5720 RH
Informations légales
Eppendorf is a registered trademark of Eppendorf AG