Principaux documents
EP022639293
Lid for Eppendorf(R) A-4-38 Rotor
aerosol-tight, pack of 2 ea
About This Item
Code UNSPSC :
41103913
Caractéristiques
aerosol-tight
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 022639293 (NA)
Technique(s)
centrifugation: suitable
Compatibilité
for use with 5702 R
for use with 5702 RH
for use with 5702
Informations légales
Eppendorf is a registered trademark of Eppendorf AG