Principaux documents
EPM1289-0700
Eppendorf® Microplate Rack
stack three deepwell or nine standard plates
About This Item
Code UNSPSC :
41122800
Caractéristiques
Holds 3 x deepwell plates
Holds 9 x plates
Fabricant/nom de marque
Eppendorf® M1289-0700 (Global)
Description générale
Microplate racks for Eppendorf incubated shakers.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG