Principaux documents
EP5895121004
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x750, aerosol tight, without buckets
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 4 x 750 mL round buckets ((aerosol-tight compatable) inc plate carrier)
to hold 4 x plate buckets
aerosol tight
swing-bucket rotor
without buckets
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5895121004 (Global)
Paramètres
4,800 × g max. RCF
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5920R
Description générale
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG