Principaux documents
EPM1289-0040
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
holds 23 tubes (18-21 mm diam.)
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.73
Caractéristiques
holds 23 tubes (18-21 mm diam.)
Fabricant/nom de marque
Eppendorf® M1289-0040 (Global)
Taille
360x80x129 mm
Description générale
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG