Principaux documents
EPM1282-9925
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 1 L Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum (heavy duty)
About This Item
Code UNSPSC :
41103816
Nomenclature NACRES :
NB.73
Matériaux
aluminum (heavy duty)
Caractéristiques
dedicated
for 1 L Erlenmeyer flasks
non-Interchangeable
Fabricant/nom de marque
Eppendorf® M1282-9925 (Global)
Capacité
(15 x 1 L Erlenmeyer flasks)
Catégories apparentées
Description générale
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
heavy duty platform for Eppendorf New Brunswick Innova 44/44R Incubator Shakers to hold 15 x 1 L Erlenmeyer flasks in dedicated, non-Interchangeable clamps.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG