Principaux documents
EPM1250-9700
Sticky Pads and Accessories
sticky pad, for all Eppendorf® shaker models, size 20 cm (8 in.) × 20 cm (8 in.)
Synonyme(s) :
Quick Change Platform Kit
About This Item
Code UNSPSC :
41103800
Nomenclature NACRES :
NB.73
Caractéristiques
sticky pad
Fabricant/nom de marque
Eppendorf® M1250-9700 (Global)
Taille
20 cm (8 in.) × 20 cm (8 in.)
Description générale
  • Sticky pads are ideal for securing culture ware (flasks, bottles, dishes) to shaker platforms that cannot be held with conventional flask clamps.
  • Reusable sticky pads can be removed easily from the surface or repositioned.
  • Used to cover the entire platform surface.
  • Easily washable with a mild detergent and water.
  • Best suited for the non-painted, anodized aluminum surface.
  • Compatible with universal platforms of New Brunswick S41i and Innova S44i.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
New Brunswick is a trademark of Eppendorf AG