Principaux documents
CLS70373160
Pyrex® Milk Dilution bottle with screw cap
capacity 160 mL, wide mouth, graduated
Synonyme(s) :
Pyrex® glass bottles
About This Item
Code UNSPSC :
41121800
Caractéristiques
soda lime glass
Conditionnement
pkg of 12 ea
case of 48 ea
Fabricant/nom de marque
Corning 70373-160
Capacité
160 mL
Informations légales
PYREX is a registered trademark of Corning, Inc.