Principaux documents
ADS41042-EP
Adapter for Eppendorf® S-4-104
holds 24 x 1.5 - 2 mL vessels, for IsoRack, pack of 2 ea
About This Item
Code UNSPSC :
41103911
Caractéristiques
holds 24 x 1.5 - 2 mL vessels
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 022638998 (NA)
Technique(s)
centrifugation: suitable
Compatibilité
for use with MTP/Flex buckets (Plate buckets)
Description générale
Tube adapters for Eppendorf rotor S-4-104.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG