Principaux documents
YY1412221
Millipore
Cartridge Housing Stainless Steel Wall Bracket Set
About This Item
Code UNSPSC :
40161525
eCl@ss :
32031603
Matériaux
stainless steel
Niveau de qualité
Diamètre
4 in.
Compatibilité
for use with Series 2000 Sanitary Cartridge Housings