Principaux documents
Z678619
2mag MIX 15 multi position stirrer
AC/DC input 100 / 240 V AC, US 2-pin plug
Synonyme(s) :
induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
stainless steel plate
Caractéristiques
positions 15
Conditionnement
pack of 1 ea
Fabricant/nom de marque
2mag 30015
Paramètres
100-2000 rpm speed
Taille de la plaque, larg. × Long.
× 245 mm × 400 mm
Volume de l'agitateur
3000 mL
Description générale
Ultra-flat magnetic stirrer with 15 stirring positions, up to 3,000 mL stirring volume, suitable for 15 x 250 mL beaker glasses.
Powerful 15 stirring positions, 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept, extremely wide speed range of 100 to 2,000 rpm, 100% synchronized speed, jerk-free stirring even at low speeds, 4-step power settings of the stirrer, high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer.