Principaux documents
Z693472
2mag MIX 15 eco multi-position magnetic stirrer
AC/DC input 110 V AC
Synonyme(s) :
laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
stainless steel plate
Conditionnement
pack of 1 ea
Fabricant/nom de marque
2mag 30115
Paramètres
120 to 1200 rpm speed (stirring)
Plaque/Plateau, l × L
240 mm × 400 mm
Volume de l'agitateur
1-1500 mL , per stirring position
Description générale
Ultra flat, wear-free magnetic stirrer with 15 stirring positions, up to 1,500 mL stirring volume suitable for 15 x 250 mL beaker glasses.
Price-optimized magnetic stirrer with 15 stirring positions, 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions, speed range from 120 to 1,200 rpm, 100% synchronized speeds, jerk-free stirring even at low speeds.
Clear digital display for stirrer speed settings, SoftStart procedure for reliable catching/centering and safe acceleration of the stirring bar. Automatic memory function to save the last settings, outstanding long-life cycle of the hermetically sealed drive system, resistant stainless steel housing for easy and fast cleaning.