Z220337
Aldrich® Schlenk-type female joint cap
female joint: ST/NS 29/32
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Produits recommandés
Étiquette/Marqueur
J
K
Joint
female joint: ST/NS29/32
Description générale
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Liaison
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC