Principaux documents
Z224170
Sigma-Aldrich
Chem-FLEX tubing clamp
for tubing o.d., 23/64 ‑ 23/64 in.
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.35
Description
For tubing CHEM-FLEX 106, 125
Caractéristiques
for tubing o.d. 23/64 ‑ 23/64 in.
Description générale
Noncorroding, lightweight nylon clamps secure CHEM-FLEX tubing to transfer needles and fittings.
Informations légales
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC