Principaux documents
Z176052
Sigma-Aldrich
Lecture-bottle valve
CGA Inlet for 180M/110F, 316 stainless steel
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
316 stainless steel
Raccords
CGA Inlet for 180M/110F
Description générale
Diffusion resistant, diaphragm seal, packless on/off valve. 3/8 in. NPTM valve thread. CGA 180M inlet/110F outlet connection.