Principaux documents
Z146706
Sigma-Aldrich
Aldrich® non-corrosive lecture-bottle gas regulator
size, CGA Inlet size 180
Synonyme(s) :
Gas control valve
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Plage de mesure
0-90 psig
Taille de l'entrée CGA
180
Taille
Description générale
  • Single-stage
  • Inlet: CGA 180
  • Outlet: 1/4" male NPTF
  • Max. inlet pressure: 3,000 psig
  • Temp. range: -40 to 140°F
  • Compact chrome-plated brass body
  • Lecture-bottle gas regulators include a PTFE sealing washer
  • Order spare PTFE sealing washers separately
Autres remarques
product is not intended for medical use
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC