Principaux documents
Z156566
Sigma-Aldrich
Aldrich® mineral-oil bubbler
threaded vacuum stopcock
Synonyme(s) :
bubblers, scientific bubblers
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Description générale
With threaded vacuum stopcock and serrated inlet/outlet tubes. For controlling and monitoring gas flow at low (<2 psi) pressures.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC