Principaux documents
SYNB262914
Synthware bubbler with PTFE stopcock
top joint: ST/NS 29/42, bottom joint: ST/NS 14/20
Synonyme(s) :
Synthware B262914, mineral oil bubbler
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS14/20
joint: ST/NS top29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware B262914
Catégories apparentées
Description générale
With small ball joints to prevent mineral oil being sucked back under sudden negative pressure in the system.
Informations légales
Synthware is a trademark of Kemtech America Inc.