SYNS630012
Synthware™ low hold up high vacuum valve with protected o-rings
size 0-12 mm
Synonyme(s) :
Synthware S630012
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Produits recommandés
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S630012
Catégories apparentées
Description générale
With two 9 mm O.D. sidearms bent to minimize hold up of liquids, Viton® O-ring on PTFE shaft are protected with precision PTFE wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal. This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company