Principaux documents
BAF201971515
SP Bel-Art® Safe-Lab Hollow PTFE Stoppers
for for 24/40 Tapered Joints
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Bel-Art F20197-1515
Raccords
for for 24/40 Tapered Joints
Description générale
Use these hollow PTFE stoppers for boiling and distilling flasks with ground joints.
Caractéristiques et avantages
  • Easily removed from standard tapered glassware by turning the red extraction nut to apply a safe, gentle and even force against the mouth of the vessel
  • Helps eliminate accidents and breakage
  • Autoclave safe at 121ºC (250ºF)
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP