Principaux documents
SYNM131514
Synthware #15 o-ring joint to inner joint adapter
joint: ST/NS 14/20
Synonyme(s) :
Synthware M131514, glass joint adapter
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware M131514
Catégories apparentées
Description générale
Synthware adapter with #15 O-Ring and a standard inner joint (14/20) to inner ground glass.
Informations légales
Synthware is a trademark of Kemtech America Inc.