Principaux documents
SYNF414251
Synthware two neck round bottom flask with angled side neck
250 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Synonyme(s) :
Synthware F414251, 2-neck round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS24/40
side joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F414251
Capacité
250 mL
250 mL
Description générale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.