Principaux documents
Z329398
BRAND® magnetic stirring bar, PTFE, triangular
L 55 mm, bar diam. 14 mm
Synonyme(s) :
PTFE, Stirring rods
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
triangular PTFE-coated
Conditionnement
pack of 10
Fabricant/nom de marque
BRAND 137822
Longueur
55 mm
Diam. de la barre
14 mm
Description générale
Wedge shaped. Ideal for dissolving solids and mixing of sediments due to scraping action. Angular surface creates high turbulence even at low speeds.
Autres remarques
BR137822 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG