Principaux documents
CLS982522
Pyrex® culture tubes with screw-cap
O.D. × L 22 mm × 175 mm
Synonyme(s) :
Corning® 9825
About This Item
Code UNSPSC :
41121706
Matériaux
cap (Rubber Liner)
round bottom tube
Conditionnement
case of 192 ea
pack of 48 ea
Fabricant/nom de marque
Corning 9825-22
D.E. × L
22 mm × 175 mm
Description générale
These Pyrex culture tubes are supplied with deep-form 18-415 GPI threaded phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of high temperatures and steam in autoclaving and are provided with a white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.