Principaux documents
CLS410050
Pyrex® flat-bottom boiling flask, short neck with 24/40 joint
capacity 50 mL
Synonyme(s) :
boiling flask, flask glass, flat bottom flasks, Corning® 4100
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
flat bottom flask
short
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Corning 4100-50
Capacité
50 mL
Taille du col
(1)
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.