Principaux documents
BMSZ32615A5
Benchmark Adapter pack for 5ml tubes in Sprint 8 Plus rotors
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
SB.56
Fabricant/nom de marque
Benchmark Scientific Z326-15-A5
Description générale
Adapter for 5ml tubes in Sprint 8 Plus Rotor
Application
Centrifugation of clinical samples including urine and blood. Ideal for production of PPP and PRP.