Principaux documents
BAF182060000
SP Bel-Art® Double Burette Clamp
for 0.5 in. Support Rod
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
Caractéristiques
to hold Double
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18206-0000
Description générale
Non-corroding clamps hold burettes firmly in place.
Caractéristiques et avantages
  • Soft rubber pads hold burette without scratching
  • Clamp fits standard 12.5mm (½") support rod and includes an adapter to fit to narrower 9.5mm (⅜") rod (support rod not included)
  • Made of chemical resistant polypropylene; autoclavable at 121°C (250°F)
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP