HUMU-210K-PMX
MILLIPLEX® Humanized Mouse Panel 41-plex Premixed and Bulk-Premixed Kit

Simultaneously analyze 41 analytes in a bead-based assay using Luminex® technology, in humanized mouse serum, plasma, and cell culture supernatants
Synonym(s):
Luminex® humanized mouse assay, Millipore humanized mouse panel, humanized mouse multiplex kit
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41116126
Recommended Products
Quality Level
quality
200
species reactivity
mouse, (human)
manufacturer/tradename
Milliplex®
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® xMAP® technology)
shipped in
wet ice
storage temp.
2-8°C
General description
Humanized mouse research has emerged as a pivotal tool in biomedical science, bridging the gap between preclinical studies and human disease. By engrafting human tissues, cells, or genes into immunocompromised mice, researchers can create models that more accurately mimic human immune responses and disease mechanisms. This innovative approach enhances the relevance of immunoassays, which are critical for evaluating immune function, disease progression, and therapeutic efficacy. As the demand for more predictive models in drug development and disease research grows, humanized mice are proving invaluable for advancing our understanding of human biology and improving therapeutic strategies.
Application
MILLIPLEX® Qualified assays undergo rigourous assay development, verification, and Quality Control testing to achieve optimal performance. Simultaneously analyze 41 analytes in humanized mouse serum, plasma, and cell culture supernatants.
Human-specific analytes included: G-CSF, GM-CSF, IFNɣ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-12p40, IL-12p70, IL-15, IL-17A, IL-18, IL-10, M-CSF, MCP-1, MIG, IP-10, TNFα Mouse-specific analytes included: G-CSF, GM-CSF, IFNɣ, IL-1β, IL-2, IL-3, IL-5, IL-6, KC, IL-12p40, IL-12p70, IL-17A, IL-18, M-CSF, MCP-1, MIG, IP-10, TNFα
Assay CharacteristicsRefer to kit protocol for assay cross-reactivity, sensitivity, precision, and accuracy.
Human-specific analytes included: G-CSF, GM-CSF, IFNɣ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-12p40, IL-12p70, IL-15, IL-17A, IL-18, IL-10, M-CSF, MCP-1, MIG, IP-10, TNFα Mouse-specific analytes included: G-CSF, GM-CSF, IFNɣ, IL-1β, IL-2, IL-3, IL-5, IL-6, KC, IL-12p40, IL-12p70, IL-17A, IL-18, M-CSF, MCP-1, MIG, IP-10, TNFα
Assay CharacteristicsRefer to kit protocol for assay cross-reactivity, sensitivity, precision, and accuracy.
Features and Benefits
- Exclusive Market Offering: The onlyhumanized mouse multiplex panel available on the market. Gain exclusive accessto advanced research capabilities that provide unique insights, enhancing thevalue of your studies.
- Hassle-Free Premixed Bead Solutions: Premixedbeads simplify your assay setup by reducing preparation time.
- Simultaneous Multi-analyte Analysis: Streamlinesthe workflow by reducing the number of individual tests required, saving timeand resources while providing comprehensive data.
- Minimal Sample Volume Requirement: Requiresonly 10 µL of sample volume per well. Economical use of precious samples,allowing for more experiments or tests to be conducted with limited sampleavailability.
- Proven Reliability and Accuracy: Ensurereproducibility and confidence in your results, which is critical for yourresearch and pre-clinical applications.
- Ready-to-use reagents: Reducespreparation time and complexity, allowing you to focus on your experimentsrather than setup.
Storage and Stability
Recommended storage for kit components is 2-8˚C
Legal Information
Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service