Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HUMU-210K

MILLIPLEX® Humanized Mouse Panel 41-plex

new

Simultaneously analyze up to 41 analytes in a bead-based assay using Luminex® technology, in humanized mouse serum, plasma, and cell culture supernatants

Synonym(s):

Luminex® humanized mouse assay, Millipore humanized mouse panel, humanized mouse multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116126

Quality Level

quality

200

species reactivity

human, mouse

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP® technology)

shipped in

wet ice

storage temp.

2-8°C

General description

Humanized mouse research has emerged as a pivotal tool in biomedical science, bridging the gap between preclinical studies and human disease. By engrafting human tissues, cells, or genes into immunocompromised mice, researchers can create models that more accurately mimic human immune responses and disease mechanisms. This innovative approach enhances the relevance of immunoassays, which are critical for evaluating immune function, disease progression, and therapeutic efficacy. As the demand for more predictive models in drug development and disease research grows, humanized mice are proving invaluable for advancing our understanding of human biology and improving therapeutic strategies.

Application

MILLIPLEX® Qualified assays undergo rigourous assay development, verification, and Quality Control testing to achieve optimal performance. Simultaneously analyze up to 41 analytes in humanized mouse serum, plasma, and cell culture supernatants.

Human-specific analytes included: G-CSF, GM-CSF, IFNɣ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-12p40, IL-12p70, IL-15, IL-17A, IL-18, IL-10, M-CSF, MCP-1, MIG, IP-10, TNFα

Mouse-specific analytes included: G-CSF, GM-CSF, IFNɣ, IL-1β, IL-2, IL-3, IL-5, IL-6, KC, IL-12p40, IL-12p70, IL-17A, IL-18, M-CSF, MCP-1, MIG, IP-10, TNFα




Assay CharacteristicsRefer to kit protocol for assay cross-reactivity, sensitivity, precision, and accuracy.

Features and Benefits

  • Exclusive Market Offering: The only humanized mouse multiplex panel available on the market. Gain exclusive access to advanced research capabilities that provide unique insights, enhancing the value of your studies.
  • Flexible Panel Options: Tailor this assay to meet your specific research needs, enhancing flexibility and efficiency in experimental design.
  • Simultaneous Multi-analyte Analysis: Streamlines the workflow by reducing the number of individual tests required, saving time and resources while providing comprehensive data.
  • Minimal Sample Volume Requirement: Requires only 10 µL of sample volume per well. Economical use of precious samples, allowing for more experiments or tests to be conducted with limited sample availability.
  • Proven Reliability and Accuracy: Ensure reproducibility and confidence in your results, which is critical for your research and pre-clinical applications.
  • Ready-to-use reagents: Reduces preparation time and complexity, allowing you to focus on your experiments rather than setup.

Storage and Stability

Recommended storage for kit components is 2-8˚C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service