Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MCYT1-190K

Millipore

MILLIPLEX® Mouse Cytokine Expansion Panel 1 68-Plex

new

Simultaneously analyze up to 68 cytokines, chemokines, and growth factors with bead-based assays using Luminex® technology, in mouse serum, plasma, and cell culture supernatants

Synonym(s):

Luminex® mouse cytokine immunoassay, Millipore mouse cytokine panel, mouse cytokine multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116126

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

detection method

fluorometric (Luminex® xMAP® technology)

shipped in

wet ice

storage temp.

2-8°C

Application

MILLIPLEX® Qualified assays undergo rigorous assay development, verification, and Quality Control testing to achieve optimal performance. Simultaneously analyze up to 68 analytes in mouse serum, plasma, and cell culture supernatants.

Analytes included: BAFF, Betacellulin, CHI3L1, CXCL16, EGF, Eotaxin/CCL11, Erythropoietin, Exodus-2/CCL21, FGF-2, FLT3L, Fractalkine/CX3CL1, G-CSF, GDF-15/MIC-1, GM-CSF, Granzyme B, IFNα, IFNβ, IFNγ, IL-10, IL-11, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-16, IL-17A, IL-17F, IL-18, IL-1α, IL-1β, IL-2, IL-20, IL-22, IL-28B/IFNλ3, IL-3, IL-31, IL-33/NF-HEV (mature), IL-4, IL-5, IL-6, IL-7, IL-9, IP-10/CXCL10, KC/GROα/CXCL1, LIF, LIX/CXCL5, MCP-1/CCL2, MCP-2/CCL8, MCP-3/CCL7, MCP-5/CCL12, M-CSF, MDC/CCL22, MIG/CXCL9, MIP-1α/CCL3, MIP-1β/CCL4, MIP-2/CXCL2, MIP-3α/CCL20, MIP-3β/CCL19, RANTES/CCL5, sCD137/4-1BB/TNFRSF9, sFas/TNFRSF6, sFasL, sICAM-1, sRAGE, TARC/CCL17, TECK/CCL25, TNFα, and VEGF-A.

The following analytes in this kit are not compatible with the MAGPIX® System: sFasL, IL-16, sFas, IL-20, sCD137, sICAM-1, CXCL16, sRAGE, CCL25/TECK, BAFF, GDF-15, FLT3L, CHI3L1, MCP-2, MCP-3, EPO, Fracktalkine, IL-31, and Betacellulin

Assay Characteristics

Refer to kit protocol for assay cross-reactivity, sensitivity, precision, and accuracy.

Features and Benefits

  • Fully Configurable Multiplex Assay: Design your own panel by selecting up to 68 analytes, enabling tailored cytokine and chemokine profiling that meets your specific research needs in biomarke discovery.
  • Efficient Sample Management: Analyze multiple targets with just 25 µL/well (1:2 diluted), conserving samples and maximizing data from limited material.
  • Adaptable incubation options: Opt for same-day or overnight incubation to seamlessly integrate into your workflow, enhancing productivity.
  • Consistent and Reliable Data: Experience proven lot-to-lot consistency for trustworthy results, vital for regulatory compliance and reproducibility in analytical R&D.
  • Streamlined Assay Setup: Ready-to-use reagents and comprehensive kit components, including Serum Matrix and Quality Controls, simplify preparation and reduce the risk of errors.

Storage and Stability

Recommended storage for kit components is 2-8˚C

Legal Information

Luminex is a registered trademark of Luminex Corp
MAGPIX is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service